1-(Cyclopropylcarbonyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049313-500MG 049313-2.5G
101887-156EA 364.62 USD
101887-156 101887-158
1-(Cyclopropylcarbonyl)-3-piperidinecarboxylic acid
1-(Cyclopropylcarbonyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MDL Number: MFCD09040653
CAS Number: 926198-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR