2-(3-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049225-500MG 049225-2.5G
101886-804EA 364.62 USD
101886-804 101886-806
2-(3-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride
2-(3-Piperidinyl)ethyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13559760
CAS Number: 1220021-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR