2-(4-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Supplier: Matrix Scientific

049203-500MG 049203-2.5G
101886-716EA 364.62 USD
101886-716 101886-718
2-(4-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride
2-(4-Piperidinyl)ethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₄ClNO₃
MDL Number: MFCD13559746
CAS Number: 1219956-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR