N-(4-Aminophenyl)tetrahydro-2-furancarboxamide
Supplier: Matrix Scientific
049017-500MG
049017-2.5G
101885-972EA
364.62
USD
101885-972
101885-974
N-(4-Aminophenyl)tetrahydro-2-furancarboxamide
N-(4-Aminophenyl)tetrahydro-2-furancarboxamide
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ |
MDL Number:
MFCD09045362 CAS Number: 926223-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...