N-(5-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049014-500MG 049014-2.5G
101885-960EA 364.62 USD
101885-960 101885-962
N-(5-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide
N-(5-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD09048726
CAS Number: 926249-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR