Ethyl-5-bromo-4,6-dichloronicotinate ≥95%

Supplier: Matrix Scientific

048688-500MG 048688-1G
101884-876EA 443.08 USD
101884-876 101884-878
Ethyl-5-bromo-4,6-dichloronicotinate ≥95%
Ethyl-5-bromo-4,6-dichloronicotinate

Hazard: Irritant

Formula: C₈H₆BrCl₂NO₂
Melting Pt: 35…37 °C
MDL Number: MFCD13193100
CAS Number: 1192263-86-7

Order Now

Specification Test Results

Melting point 35-37°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR