2,2-Dimethyl-5-vinyl-1,3-dioxan-5-ol ≥95%

Supplier: Matrix Scientific

048602-500MG 048602-1G 048602-5G
101884-544EA 747.69 USD
101884-544 101884-546 101884-548
2,2-Dimethyl-5-vinyl-1,3-dioxan-5-ol ≥95%
2,2-Dimethyl-5-vinyl-1,3-dioxan-5-ol

Hazard: Irritant

Formula: C₈H₁₄O₃
MDL Number: MFCD12922679
CAS Number: 933791-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR