4-Bromopicolinic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Bromopyridine-2-carboxylic acid

048494-1G 048494-5G
101884-076EA 26.15 USD
101884-076 101884-078
4-Bromopicolinic acid ≥98% for synthesis
4-Bromopicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄BrNO₂
Melting Pt: 172…173 °C
MDL Number: MFCD03426665
CAS Number: 30766-03-1

Order Now

Specification Test Results

Melting point 172-173°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR