5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Dimethoxybenzo[b]thiophene-2-carboxylic acid

048280-500MG 048280-1G
101883-586EA 556.92 USD
101883-586 101883-588
5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid ≥95%
5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀O₄S
MDL Number: MFCD08275132
CAS Number: 23046-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR