5-Morpholino-2-nitrobenzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

048224-500MG 048224-1G
101883-360EA 556.92 USD
101883-360 101883-362
5-Morpholino-2-nitrobenzenecarbohydrazide ≥95%
5-Morpholino-2-nitrobenzenecarbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₄N₄O₄
Melting Pt: 211…213 °C
MDL Number: MFCD11841057
CAS Number: 1135283-85-0

Order Now

Specification Test Results

Melting point 211-213°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR