Methyl-5-bromo-2-piperidinonicotinate ≥95%

Supplier: Matrix Scientific

048173-500MG 048173-1G 048173-5G
101883-156EA 556.92 USD
101883-156 101883-158 101883-160
Methyl-5-bromo-2-piperidinonicotinate ≥95%
Methyl-5-bromo-2-piperidinonicotinate

Hazard: Irritant

Formula: C₁₂H₁₅BrN₂O₂
MW: 299.17 g/mol
MDL Number: MFCD09972267
CAS Number: 1017783-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR