Ethyl-2-(tetrahydro-2H-pyran-2-ylamino)-1,3-thiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

047547-500MG 047547-1G 047547-5G
101880-530EA 278.46 USD
101880-530 101880-532 101880-534
Ethyl-2-(tetrahydro-2H-pyran-2-ylamino)-1,3-thiazole-5-carboxylate ≥95%
Ethyl-2-(tetrahydro-2H-pyran-2-ylamino)-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃S
Melting Pt: 129…131 °C
MDL Number: MFCD11553044
CAS Number: 1029088-17-2

Order Now

Specification Test Results

Melting point 129-131°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR