[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl](morpholino)methanone ≥95%

Supplier: Matrix Scientific

047450-1G
101880-048EA 467.69 USD
101880-048
[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl](morpholino)methanone ≥95%
[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl](morpholino)methanone

Hazard: Irritant

Formula: C₁₅H₁₇BrN₄O₂
Melting Pt: 166 °C
MDL Number: MFCD09607956
CAS Number: 952182-52-4

Order Now

Specification Test Results

Melting point 166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR