7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

047324-500MG
101879-502EA 166.15 USD
101879-502
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅F₃O₂S
Melting Pt: 191…195 °C
MDL Number: MFCD09027110
CAS Number: 550998-66-8

Order Now

Specification Test Results

Melting point 191-195°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR