Ethyl-1,6-dimethyl-4-(3-methylphenyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047139-500MG 047139-1G 047139-5G
101878-612EA 181.54 USD
101878-612 101878-614 101878-616
Ethyl-1,6-dimethyl-4-(3-methylphenyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-1,6-dimethyl-4-(3-methylphenyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₃
MW: 288.35 g/mol
Melting Pt: 126…129 °C
MDL Number: MFCD12025937
CAS Number: 1135283-41-8

Order Now

Specification Test Results

Melting point 126-129°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR