Ethyl-4-(4-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047136-500MG 047136-1G 047136-5G
101878-594EA 181.54 USD
101878-594 101878-596 101878-598
Ethyl-4-(4-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(4-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₄
MW: 304.35 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00551253
CAS Number: 219814-75-2

Order Now

Specification Test Results

Melting point 110-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR