1-[(2-Nitrophenyl)sulfonyl]piperazine ≥95%
Supplier: Matrix Scientific
047101-500MG
047101-1G
101878-464EA
132.31
USD
101878-464
101878-466
1-[(2-Nitrophenyl)sulfonyl]piperazine ≥95%
1-[(2-Nitrophenyl)sulfonyl]piperazine
Hazard: Irritant
Formula:
C₁₀H₁₃N₃O₄S Melting Pt: 146…148 °C |
MDL Number:
MFCD00227564 CAS Number: 301331-16-8 |
Specification Test Results
Melting point | 146-148°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...