Methyl-2-fluoro-4-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-fluoro-4-nitrobenzoate

046852-5G 046852-25G
101877-278EA 80 USD
101877-278 101877-280
Methyl-2-fluoro-4-nitrobenzoate ≥95%
Methyl 2-fluoro-4-nitrobenzoate

Hazard: Irritant

Formula: C₈H₆FNO₄
Melting Pt: 73…75 °C
MDL Number: MFCD08444028
CAS Number: 392-09-6

Order Now

Specification Test Results

Melting point 73-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR