Ethyl-8-amino-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

046759-500MG 046759-1G
101876-896EA 461.54 USD
101876-896 101876-898
Ethyl-8-amino-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 8-amino-2-methylimidazo[1,2-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
Melting Pt: 150…152 °C
MDL Number: MFCD10758082
CAS Number: 185133-90-8

Order Now

Specification Test Results

Melting point 150-152°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR