2-[1-Benzyl-4-(hydroxymethyl)tetrahydro-1H-pyrrol-3-yl]benzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[1-Benzyl-4-(hydroxymethyl)pyrrolidin-3-yl]phenol

046752-500MG 046752-1G
101876-866EA 921.54 USD
101876-866 101876-868
2-[1-Benzyl-4-(hydroxymethyl)tetrahydro-1H-pyrrol-3-yl]benzenol ≥95%
2-[1-Benzyl-4-(hydroxymethyl)tetrahydro-1H-pyrrol-3-yl]benzenol

Hazard: Irritant

Formula: C₁₈H₂₁NO₂
MDL Number: MFCD10758080
CAS Number: 1033463-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR