tert-Butyl-4-[3-(1H-pyrazol-5-yl)phenethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[3-(1H-Pyrazol-5-yl)phenethyl]piperazine, N1-BOC protected

046718-500MG 046718-1G
101876-716EA 747.69 USD
101876-716 101876-718
tert-Butyl-4-[3-(1H-pyrazol-5-yl)phenethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[3-(1H-pyrazol-5-yl)phenethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₂₀H₂₈N₄O₂
Melting Pt: 88…90 °C
MDL Number: MFCD08444013
CAS Number: 1186648-43-0

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR