2-Methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046520-500MG 046520-1G 046520-5G
101875-862EA 375.38 USD
101875-862 101875-864 101875-866
2-Methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%
2-Methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₆
Melting Pt: 173 °C
MDL Number: MFCD08443984
CAS Number: 154365-43-2

Order Now

Specification Test Results

Melting point 173°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR