4'-Chlorobiphenyl-2-carboxylic acid ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 4'-Chloro-[1,1'-biphenyl]-2-carboxylic acid, 4'-Chloro[1,1'-biphenyl]-2-carboxylic acid

046491-500MG 046491-1G
101875-712EA 375.38 USD
101875-712 101875-714
4'-Chlorobiphenyl-2-carboxylic acid ≥95% for synthesis
4'-Chlorobiphenyl-2-carboxylic acid

Hazard code(s): Irritant

Formula: C₁₃H₉ClO₂
Melting Pt: 167…169 °C
MDL Number: MFCD03426504
CAS Number: 7079-15-4

Order Now

Specification Test Results

Melting point 167-169°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR