2,4-Dichloronicotinc acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloropyridine-3-carboxylic acid, 2,4-Dichloro-3-pyridinecarboxylic acid, 2,4-Dichloronicotinic acid

046443-1G 046443-5G
101875-518EA 29.23 USD
101875-518 101875-520
2,4-Dichloronicotinc acid ≥95%
2,4-Dichloronicotinc acid

Hazard: Irritant

Formula: C₆H₃Cl₂NO₂
Melting Pt: 166…167 °C
MDL Number: MFCD06796292
CAS Number: 262423-77-8

Order Now

Specification Test Results

Melting point 166-167°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR