3'-Nitro[1,1'-biphenyl]-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

046433-500MG 046433-1G 046433-5G
101875-470EA 278.46 USD
101875-470 101875-472 101875-474
3'-Nitro[1,1'-biphenyl]-4-carboxylic acid ≥95%
3'-Nitro[1,1'-biphenyl]-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉NO₄
Melting Pt: >300 °C
MDL Number: MFCD01846432
CAS Number: 5737-85-9

Order Now

Specification Test Results

Melting point 300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR