4-Chloro-2-phenyl-6-piperidinopyrimidine ≥95%

Supplier: Matrix Scientific

046188-500MG 046188-1G 046188-5G
101874-594EA 375.38 USD
101874-594 101874-596 101874-598
4-Chloro-2-phenyl-6-piperidinopyrimidine ≥95%
4-Chloro-2-phenyl-6-piperidinopyrimidine

Hazard: Irritant

Formula: C₁₅H₁₆ClN₃
Melting Pt: 124…126 °C
MDL Number: MFCD00754197
CAS Number: 574708-35-3

Order Now

Specification Test Results

Melting point 124-126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR