4-[(3,4-Difluorobenzyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

046182-500MG 046182-1G 046182-5G
101874-562EA 181.54 USD
101874-562 101874-564 101874-566
4-[(3,4-Difluorobenzyl)oxy]benzenecarbaldehyde ≥95%
4-[(3,4-Difluorobenzyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀F₂O₂
Melting Pt: 88…90 °C
MDL Number: MFCD09972208
CAS Number: 1017782-47-6

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR