2-(4-Chlorophenyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

046011-500MG 046011-1G 046011-5G
101873-726EA 601.54 USD
101873-726 101873-728 101873-730
2-(4-Chlorophenyl)nicotinaldehyde ≥95%
2-(4-Chlorophenyl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈ClNO
Melting Pt: 91…94 °C
MDL Number: MFCD06659614
CAS Number: 885950-19-6

Order Now

Specification Test Results

Melting point 91-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR