2-Nitro-5-(1-pyrrolidinyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

045922-500MG 045922-1G 045922-5G
101873-274EA 375.38 USD
101873-274 101873-276 101873-278
2-Nitro-5-(1-pyrrolidinyl)benzenecarboxamide ≥95%
2-Nitro-5-(1-pyrrolidinyl)benzenecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₃
Melting Pt: 246…248 °C
MDL Number: MFCD03791248
CAS Number: 299897-90-8

Order Now

Specification Test Results

Melting point 246-248°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR