Ethyl-5-aminobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
045750-500MG
045750-1G
045750-5G
101872-416EA
150.77
USD
101872-416
101872-418
101872-420
Ethyl-5-aminobenzo[b]thiophene-2-carboxylate ≥95%
Ethyl 5-aminobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂S Melting Pt: 92…95 °C |
CAS Number:
25785-10-8 |
Specification Test Results
Melting point | 92-95°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...