tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 1-Oxa-6-azaspiro[2.5]octane, N-BOC protected

045685-1G 045685-5G 045685-25G
101872-096EA 56.92 USD
101872-096 101872-098 101872-100
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥95%
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
Melting Pt: 45…48 °C
MDL Number: MFCD07779385
CAS Number: 147804-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR