(6-Chloroimidazo[2,1-b][1,3]thiazol-5-yl)methanol ≥95%

Supplier: Matrix Scientific

045501-500MG 045501-1G
101871-174EA 921.54 USD
101871-174 101871-176
(6-Chloroimidazo[2,1-b][1,3]thiazol-5-yl)methanol ≥95%
(6-Chloroimidazo[2,1-b][1,3]thiazol-5-yl)methanol

Hazard: Irritant

Formula: C₆H₅ClN₂OS
Melting Pt: 168…170 °C
MDL Number: MFCD02253174
CAS Number: 24918-13-6

Order Now

Specification Test Results

Melting point 168-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR