N-(4-Fluorobenzyl)urea ≥95%

Supplier: Matrix Scientific

045333-500MG 045333-1G 045333-5G
101870-362EA 181.54 USD
101870-362 101870-364 101870-366
N-(4-Fluorobenzyl)urea ≥95%
N-(4-Fluorobenzyl)urea

Hazard: Irritant

Formula: C₈H₉FN₂O
Melting Pt: 185…187 °C
MDL Number: MFCD00085086
CAS Number: 76523-24-5

Order Now

Specification Test Results

Melting point 185-187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR