5-[3-(Trifluoromethyl)phenyl]-2-pyrimidinylhydrosulfide ≥95%

Supplier: Matrix Scientific

Synonyms: 5-[3-(Trifluoromethyl)phenyl]pyrimidine-2-thiol

045288-500MG 045288-1G 045288-5G
101870-118EA 747.69 USD
101870-118 101870-120 101870-122
5-[3-(Trifluoromethyl)phenyl]-2-pyrimidinylhydrosulfide ≥95%
5-[3-(Trifluoromethyl)phenyl]-2-pyrimidinylhydrosulfide

Hazard: Irritant

Formula: C₁₁H₇F₃N₂S
Melting Pt: 262…265 °C
MDL Number: MFCD00243479
CAS Number: 344282-81-1

Order Now

Specification Test Results

Melting point 262-265°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR