3-Phenyl-2,1-benzisoxazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

045269-500MG 045269-1G 045269-5G
101870-032EA 376.92 USD
101870-032 101870-034 101870-036
3-Phenyl-2,1-benzisoxazole-5-carbaldehyde ≥95%
3-Phenyl-2,1-benzisoxazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₉NO₂
Melting Pt: 158…160 °C
MDL Number: MFCD00623237
CAS Number: 94752-06-4

Order Now

Specification Test Results

Melting point 158-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR