3-[(4-Methylbenzyl)oxy]-2-thiophenecarbohydrazide ≥95%
Supplier: Matrix Scientific
045233-500MG
045233-1G
045233-5G
101869-828EA
375.38
USD
101869-828
101869-830
101869-832
3-[(4-Methylbenzyl)oxy]-2-thiophenecarbohydrazide ≥95%
3-[(4-Methylbenzyl)oxy]-2-thiophenecarbohydrazide
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₂S MW: 262.33 g/mol Melting Pt: 134…136 °C |
MDL Number:
MFCD00215167 CAS Number: 343375-82-6 |
Specification Test Results
Melting point | 134-136°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...