Ethyl-3-(4-isopropylphenyl)acrylate ≥95%

Supplier: Matrix Scientific

045197-500MG 045197-1G 045197-5G
101869-640EA 278.46 USD
101869-640 101869-642 101869-644
Ethyl-3-(4-isopropylphenyl)acrylate ≥95%
Ethyl-3-(4-isopropylphenyl)acrylate

Hazard: Irritant

Formula: C₁₄H₁₈O₂
MDL Number: MFCD08056649
CAS Number: 32580-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR