2-Chloro-6-iodoaniline ≥95%

Supplier: Matrix Scientific

045090-500MG 045090-1G 045090-5G
101869-126EA 375.38 USD
101869-126 101869-128 101869-130
2-Chloro-6-iodoaniline ≥95%
2-Chloro-6-iodoaniline

Hazard: Irritant

Formula: C₆H₅ClIN
MW: 253.4681 g/mol
Melting Pt: 66…67 °C
MDL Number: MFCD08443948
CAS Number: 84483-28-3

Order Now

Specification Test Results

Melting point 66-67°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR