Ethyl-4-chloroquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate

045037-500MG 045037-1G 045037-5G
101868-834EA 556.92 USD
101868-834 101868-836 101868-838
Ethyl-4-chloroquinoline-3-carboxylate ≥95%
Ethyl 4-chloroquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂
Melting Pt: 46…48 °C
MDL Number: MFCD00173407
CAS Number: 13720-94-0

Order Now

Specification Test Results

Melting point 46-48°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR