6-(4-Nitrophenyl)-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

045002-500MG 045002-1G 045002-5G
101868-658EA 1109.23 USD
101868-658 101868-660 101868-662
6-(4-Nitrophenyl)-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine ≥95%
6-(4-Nitrophenyl)-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₃H₁₃N₅O₂
Melting Pt: 262…265 °C
MDL Number: MFCD00180838
CAS Number: 338791-45-0

Order Now

Specification Test Results

Melting point 262-265°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR