3-(1-Bromo-1-methylethyl)-1,5-dihydro-2,4-benzodioxepine ≥95%

Supplier: Matrix Scientific

044938-500MG 044938-1G 044938-5G
101868-322EA 556.92 USD
101868-322 101868-324 101868-326
3-(1-Bromo-1-methylethyl)-1,5-dihydro-2,4-benzodioxepine ≥95%
3-(1-Bromo-1-methylethyl)-1,5-dihydro-2,4-benzodioxepine

Hazard: Irritant

Formula: C₁₂H₁₅BrO₂
MW: 271.16 g/mol
Melting Pt: 66…69 °C
MDL Number: MFCD06659562
CAS Number: 322408-29-7

Order Now

Specification Test Results

Melting point 66-69°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR