2-[(2-Carboxyethyl)sulfanyl]nicotinic acid ≥95%

Supplier: Matrix Scientific

044857-500MG 044857-1G 044857-5G
101867-920EA 278.46 USD
101867-920 101867-922 101867-924
2-[(2-Carboxyethyl)sulfanyl]nicotinic acid ≥95%
2-[(2-Carboxyethyl)sulfanyl]nicotinic acid

Hazard: Irritant

Formula: C₉H₉NO₄S
Melting Pt: 203…205 °C
MDL Number: MFCD02250406
CAS Number: 286472-02-4

Order Now

Specification Test Results

Melting point 203-205°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR