5-(3-Thienyl)thiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: [2,3'-Bithiophene]-5-carboxylic acid

044697-500MG 044697-1G
101867-160EA 921.54 USD
101867-160 101867-162
5-(3-Thienyl)thiophene-2-carboxylic acid ≥95%
5-(3-Thienyl)thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆O₂S₂
MW: 210.27 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD05885496
CAS Number: 60141-31-3

Order Now

Specification Test Results

Melting point 188-190°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR