2-Fluoro-4-(3-thienyl)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

044687-500MG 044687-1G 044687-5G
101867-104EA 556.92 USD
101867-104 101867-106 101867-108
2-Fluoro-4-(3-thienyl)benzenecarbonitrile ≥95%
2-Fluoro-4-(3-thienyl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₁H₆FNS
Melting Pt: 84…87 °C
MDL Number: MFCD07698592
CAS Number: 886361-69-9

Order Now

Specification Test Results

Melting point 84-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR