Ethyl-1-(2-bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

044670-500MG 044670-1G 044670-5G
101867-010EA 278.46 USD
101867-010 101867-012 101867-014
Ethyl-1-(2-bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(2-bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₅BrN₂O₂
MW: 323.2 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD07021454
CAS Number: 957044-07-4

Order Now

Specification Test Results

Melting point 86-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR