3-(Dimethylamino)-1,2-bis(4-methoxyphenyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

044592-500MG 044592-1G 044592-5G
101866-620EA 278.46 USD
101866-620 101866-622 101866-624
3-(Dimethylamino)-1,2-bis(4-methoxyphenyl)-2-propen-1-one ≥95%
3-(Dimethylamino)-1,2-bis(4-methoxyphenyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₉H₂₁NO₃
Melting Pt: 119…121 °C
MDL Number: MFCD03425780
CAS Number: 66521-59-3

Order Now

Specification Test Results

Melting point 119-121°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR