L-tert-Leucine methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Methyl 2-amino-3,3-dimethylbutanoate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester, H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate

044518-500MG 044518-1G 044518-5G
101866-296EA 369.23 USD
101866-296 101866-298 101866-300
L-tert-Leucine methyl ester ≥95%
L-tert-Leucine methyl ester

Purity: 95% CAS: 63038-26-6 MDL Number: MFCD00137409 Molecular Formula: C7H15NO2 Molecular Weight: 145,2 Hazard: Irritant

Formula: C₇H₁₅NO₂
MDL Number: MFCD00137409
CAS Number: 63038-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR