2-({2-[(4-Methoxybenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid ≥95%

Supplier: Matrix Scientific

044440-500MG 044440-1G 044440-5G
101865-856EA 181.54 USD
101865-856 101865-858 101865-860
2-({2-[(4-Methoxybenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid ≥95%
2-({2-[(4-Methoxybenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄S
Melting Pt: 92…94 °C
MDL Number: MFCD00202395
CAS Number: 338421-45-7

Order Now

Specification Test Results

Melting point 92-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR