5-Chloro-4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

044399-500MG 044399-1G 044399-5G
101865-634EA 461.54 USD
101865-634 101865-636 101865-638
5-Chloro-4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥95%
5-Chloro-4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₆H₃ClF₃N₃
Melting Pt: 84…86 °C
MDL Number: MFCD00140981
CAS Number: 219568-17-9

Order Now

Specification Test Results

Melting point 84-86°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR