Methyl-1-[(4-chlorophenyl)sulfonyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044222-500MG 044222-1G 044222-5G
101864-800EA 278.46 USD
101864-800 101864-802 101864-804
Methyl-1-[(4-chlorophenyl)sulfonyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[(4-chlorophenyl)sulfonyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₄S
Melting Pt: 131…133 °C
MDL Number: MFCD01045654
CAS Number: 331828-48-9

Order Now

Specification Test Results

Melting point 131-133°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR